Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichloro(Ethylenediamine)Platinum(II)

Catalog No. 244929250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-492-9250MG 250 mg
24-492-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 24-492-9250MG Supplier Sigma Aldrich Supplier No. 244929250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dichloro(Ethylenediamine)Platinum(II)

Specifications

CAS 14096-51-6
Molecular Formula C2H8Cl2N2Pt
Linear Formula (H2NCH2CH2NH2)PtCl2
MDL Number MFCD00000011
Quantity 250 mg
Synonym Dichloro(1,2-diaminoethane)platinum; Dichloro(1,2-ethanediamine)platinum; Platinum ethylenediamine dichloride
Molecular Weight (g/mol) 326.08 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.