Learn More
Strem, An Ascensus Company Cl2(Me5Cp)iridiumdimer98%500mg
SDP
Catalog No. 5090113121
Encompass
CAS# 12354-84-6. 500mg. Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, 98%. MFCD00075435. Molecular Weight: 796.67. Molecular Formula: [(CH3)5C5IrCl2]2. Color/form: orange xtl. Strem# 77-1060. http://www.strem.com/catalog/v/77-1060/
Catalog No. 50-901-13121 Supplier Strem, An Ascensus Company Supplier No. 771060500MG
CAS# 12354-84-6. 500mg. Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, 98%. MFCD00075435. Molecular Weight: 796.67. Molecular Formula: [(CH3)5C5IrCl2]2. Color/form: orange xtl. Strem# 77-1060. http://www.strem.com/catalog/v/77-1060/