Learn More
Strem, An Ascensus Company RhCl2(Me5Cp) dimer 99% 500mg
SDP
Catalog No. 5090112106
Encompass
CAS# 12354-85-7. 500mg. Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, 99%. MFCD00061552. Molecular Weight: 618.08. Molecular Formula: [(CH3)5C5RhCl2]2. Color/form: red xtl. Strem# 45-0195. http://www.strem.com/catalog/v/45-0195/
Catalog No. 50-901-12106 Supplier Strem, An Ascensus Company Supplier No. 450195500MG
CAS# 12354-85-7. 500mg. Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, 99%. MFCD00061552. Molecular Weight: 618.08. Molecular Formula: [(CH3)5C5RhCl2]2. Color/form: red xtl. Strem# 45-0195. http://www.strem.com/catalog/v/45-0195/