Learn More
Strem, An Ascensus Company CAS# 96503-27-4. 1g. Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer. MFCD00070449
SDP
Catalog No. 5090111924
Encompass
CAS# 96503-27-4. 1g. Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer. MFCD00070449. Molecular Weight: (307.20)n. Molecular Formula: [C5(CH3)5RuCl2]n. Color/form: red to brown pwdr. Strem# 44-0440. http://www.strem.com/catalog/v/44-0440/
Catalog No. 50-901-11924 Supplier Strem, An Ascensus Company Supplier No. 4404401G
CAS# 96503-27-4. 1g. Dichloro(pentamethylcyclopentadienyl)ruthenium(III) polymer. MFCD00070449. Molecular Weight: (307.20)n. Molecular Formula: [C5(CH3)5RuCl2]n. Color/form: red to brown pwdr. Strem# 44-0440. http://www.strem.com/catalog/v/44-0440/