Learn More
Strem, An Ascensus Company Cl2(Me5Cp)iridium dimer 98% 2g
SDP
Catalog No. 5090113122
Encompass
CAS# 12354-84-6. 2g. Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, 98%. MFCD00075435. Molecular Weight: 796.67. Molecular Formula: [(CH3)5C5IrCl2]2. Color/form: orange xtl. Strem# 77-1060. http://www.strem.com/catalog/v/77-1060/
Catalog No. 50-901-13122 Supplier Strem, An Ascensus Company Supplier No. 7710602G
CAS# 12354-84-6. 2g. Dichloro(pentamethylcyclopentadienyl)iridium(III) dimer, 98%. MFCD00075435. Molecular Weight: 796.67. Molecular Formula: [(CH3)5C5IrCl2]2. Color/form: orange xtl. Strem# 77-1060. http://www.strem.com/catalog/v/77-1060/