Learn More
Strem, An Ascensus Company CAS# 115826-95-4. 250mg. Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II), min. 98%. MFCD00075254
SDP
Supplier:  Strem, An Ascensus Company 460870250MG
Encompass
CAS# 115826-95-4. 250mg. Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]palladium(II), min. 98%. MFCD00075254. Molecular Weight: 800.00. Molecular Formula: PdCl2(C44H32P2). Color/form: orange microxtls. Strem# 46-0870. http://www.strem.com/catalog/v/46-0870/
Catalog No. NC1662352