Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetaldehyde Hydrate 90.0+%, TCI America™
SDP

Catalog No. D031225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D031225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D031225G Supplier TCI America Supplier No. D031225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16086-14-9
Molecular Formula C2H2Cl2O
MDL Number MFCD00150652
Synonym 2,2-Dichloro-1,1-ethanediol

Specifications

Melting Point 56°C
Color White
Boiling Point 121°C
Quantity 25 g
UN Number 1759
Formula Weight 112.94
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dichloroacetaldehyde Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.