Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichloroacetic Acid

Catalog No. D54702500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54702500ML 500 mL
D5470225L 2.5 L
D54702 4 x 100 mL
D54702100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. D54702500ML Supplier Sigma Aldrich Supplier No. D54702500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dichloroacetic Acid

Specifications

CAS 79-43-6
Melting Point 9°C to 11°C
Density 1.563 g/mL at 25°C
Boiling Point 194°C
Molecular Formula C2H2Cl2O2
Refractive Index n20/D 1.466
Linear Formula Cl2CHCOOH
MDL Number MFCD00004223
Quantity 500 mL
Molecular Weight (g/mol) 128.94 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.