Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetic acid, 99%

Catalog No. AAA147400E
Change view
Click to view available options
Quantity:
2500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA147400E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AAA147400E Supplier Thermo Scientific Chemicals Supplier No. A147400E

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-43-6 | C2H2Cl2O2 | 128.936 g/mol

Dichloroacetic acid salts are used as drugs due to its inhibition of the enzyme pyruvate dehydrogenase kinase. It is also employed to stem the production of lactic acid to treat lactic acidosis. It is clinically essential in children to treat lactic acidosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Miscible with water, ethanol, diethyl ether, n-octanol and acetone.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-43-6
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00004223
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
Synonym dichloroacetic acid, dichloracetic acid, dichloroethanoic acid, acetic acid, dichloro, urner's liquid, dichlorethanoic acid, bichloracetic acid, dca acid, bichloroacetic acid, dichloro-acetic acid
PubChem CID 6597
ChEBI CHEBI:36386
IUPAC Name 2,2-dichloroacetic acid
SMILES C(C(=O)O)(Cl)Cl

Specifications

Melting Point 9°C to 11°C
Density 1.564
Boiling Point 192°C to 194°C
Flash Point >110°C (230°F)
Odor Pungent
Assay Percent Range 99%
Refractive Index 1.462
Quantity 2500 g
UN Number UN1764
Beilstein 1098596
Sensitivity Hygroscopic
Merck Index 14,3050
Solubility Information Miscible with water,ethanol,diethyl ether,n-octanol and acetone.
Formula Weight 128.94
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dichloroacetic acid
Show More Show Less
Hazard Category H290-H311-H314-H351-H360-H362-H373
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P201-P202-P234-P260-P263-P264b-P270-P271-P281-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DICHLOROACETIC ACID
EINECSNumber 201-207-0
RTECSNumber AG6125000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.