Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroacetic acid, technical, Thermo Scientific™

Catalog No. AC473140025
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC473140025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC473140025 Supplier Thermo Scientific Chemicals Supplier No. 473140025
  • $719.50
    $611.65 / Each of 1

    Save $107.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 79-43-6
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.94
MDL Number 00004223
InChI Key JXTHNDFMNIQAHM-UHFFFAOYSA-N
IUPAC Name 2,2-dichloroacetic acid
SMILES OC(=O)C(Cl)Cl

Specifications

Alpha Vector DICHLOROACETICACID
Color Clear colorless to dark yellow
Flash Point >112°C
Infrared Spectrum Conforms
Assay Percent Range 98.5 to 101.5% (Titration with NaOH)
Packaging Glass Bottle
Linear Formula Cl2CHCO2H
Refractive Index 1.4650 to 1.4670 (20°C, 589 nm)
Quantity 2.5 L
Solubility Information Soluble in water; Miscible with alcohol, ether and ethanol; Soluble in acetone
Physical Form Liquid
Chemical Name or Material Dichloroacetic acid
Show More Show Less
Hazard Category Corrosion
Hazard Statement Skull and crossbones
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.