Learn More
Strem, An Ascensus Company CAS# 14220-64-5. 5g. Dichlorobis(benzonitrile)palladium(II), 99%. MFCD00013123
SDP
Catalog No. 5090112334
Encompass
CAS# 14220-64-5. 5g. Dichlorobis(benzonitrile)palladium(II), 99%. MFCD00013123. Molecular Weight: 383.55. Molecular Formula: PdCl2(C6H5CN)2. Color/form: yellow to orange pwdr. Strem# 46-0400. http://www.strem.com/catalog/v/46-0400/
Catalog No. 50-901-12334 Supplier Strem, An Ascensus Company Supplier No. 4604005G
CAS# 14220-64-5. 5g. Dichlorobis(benzonitrile)palladium(II), 99%. MFCD00013123. Molecular Weight: 383.55. Molecular Formula: PdCl2(C6H5CN)2. Color/form: yellow to orange pwdr. Strem# 46-0400. http://www.strem.com/catalog/v/46-0400/