Learn More
Medchemexpress LLC Dichlorobis(tricyclohexylphosphine)palladium(II) | 29934-17-6 | MFCD00191830 | 98.0% | 738.2 g/mol | C36H66Cl2P2Pd | 5 G
SDP
Catalog No. 5000301768
Encompass_Preferred
Dichlorobis(tricyclohexylphosphine)palladium(II) is a palladium(II) phosphine complex used as a homogeneous catalyst in organic synthesis. It is commonly employed for C-C and C-N cross-coupling reactions and for the carbonylation of chloroarenes under mild conditions, and is supplied as a solid for research use.
  • Used as a homogeneous catalyst for C-C and C-N cross-coupling reactions.
  • Effective in the carbonylation of chloroarenes under mild conditions.
  • High purity suitable for research applications.
  • Solid form for straightforward handling and storage.
Catalog No. 50-003-01768 Supplier Medchemexpress LLC Supplier No. HYW0333125G
Dichlorobis(tricyclohexylphosphine)palladium(II) is a palladium(II) phosphine complex used as a homogeneous catalyst in organic synthesis. It is commonly employed for C-C and C-N cross-coupling reactions and for the carbonylation of chloroarenes under mild conditions, and is supplied as a solid for research use.
  • Used as a homogeneous catalyst for C-C and C-N cross-coupling reactions.
  • Effective in the carbonylation of chloroarenes under mild conditions.
  • High purity suitable for research applications.
  • Solid form for straightforward handling and storage.