Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichloroborane Dioxane Complex Solution

Catalog No. 55595925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-595-925ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 55-595-925ML Supplier Sigma Aldrich Supplier No. 55595925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Hydroboration reactionsPreparation of alkenyl- and alkylboronic acidsU.S. Pat. No. 6,008,414

Specifications

CAS 22148-86-3
Density 0.976 g/mL at 25°C
Boiling Point 206°C to 207°C
Molecular Formula C10H14O
Linear Formula C6H5(CH2)2CH(OH)CH3
MDL Number MFCD03093082
Quantity 1 g
Molecular Weight (g/mol) 150.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.