Learn More
AdipoGen Dichlorocamphorylsulfonyl oxaz
SDP
Supplier:  AdipoGen CDXD0031G005
Encompass_Preferred
Chemical. CAS 127184-05-8. Formula C10H13Cl2NO3S. MW 298.19. Synthetic. Chiral oxidizing agent. Used for asymmetric synthesis as chiral catalyst. Hydroxylation reagent.
Catalog No. 50-256-1303