Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichlorodiisopropylsilane

Catalog No. 383855ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-385-5ML 5 mL
38-385-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 38-385-5ML Supplier Sigma Aldrich Supplier No. 383855ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protecting group reagent; brings two alcohols in close contact for tethering technique

Specifications

CAS 7751-38-4
Density 1.026 g/mL at 20°C
Boiling Point 66°C (27 mmHg, lit.)
Molecular Formula C6H14Cl2Si
Refractive Index n20/D 1.444
Linear Formula C6H14Cl2Si
MDL Number MFCD00054895
Quantity 5 mL
Synonym Diisopropyldichlorosilane
Molecular Weight (g/mol) 185.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.