Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichlorodimethylsilane

Catalog No. 40140500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-140-500ML 500 mL
40-140-100ML 100 mL
40-140-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 40-140-500ML Supplier Sigma Aldrich Supplier No. 40140500ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dichlorodimethylsilane

Specifications

CAS 75-78-5
Melting Point -76°C
Density 1.07 g/mL at 25°C
Boiling Point 70°C
Molecular Formula C2H6Cl2Si
Refractive Index n20/D 1.404
Linear Formula (CH3)2SiCl2
MDL Number MFCD00000491
Quantity 500 mL
Synonym DMDCS; Dimethyldichlorosilane
Molecular Weight (g/mol) 129.06 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.