Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorodimethylsilane 98.0+%, TCI America™
SDP

Catalog No. D035825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D035825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D035825ML Supplier TCI America Supplier No. D035825ML

missing translation for 'validMainframeMsg'

Specifications

CAS 75-78-5
Melting Point -76°C
Color Yellow
Boiling Point 70°C
Molecular Formula C2H6Cl2Si
Quantity 25 mL
MDL Number MFCD00000491
UN Number 1162
Synonym dimethyldichlorosilane, silane, dichlorodimethyl, dichloro dimethyl silane, inerton aw-dmcs, dichlorodimethylsilicon, dimethyl-dichlorsilan, inerton dw-dmc, repel-silan, dimethylsilane dichloride, dimethyl dichlorosilane
InChI Key LIKFHECYJZWXFJ-UHFFFAOYSA-N
SMILES C[Si](C)(Cl)Cl
IUPAC Name dichloro(dimethyl)silane
Molecular Weight (g/mol) 129.055
PubChem CID 6398
Formula Weight 129.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorodimethylsilane
Show More Show Less
EINECSNumber (2)-2041
RTECSNumber VV3150000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.