Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dichlorodiphenylsilane

Catalog No. D61504100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D61504100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D61504100G Supplier Sigma Aldrich Supplier No. D61504100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dichlorodiphenylsilane is a silane based surface modifier, and a precursor which can be used in the synthesis of silica based materials. It is used to modify the surface by providing toughness, durability and resistance to shock.The addition reaction of dichlorodiphenylsilane with diglycidyl ether of bisphenol-S yields poly(silyl ether) with pendant chloromethyl groups. {40} The coupling reaction between dichloromethylsilane and dichlorodiphenylsilane can form diorganosilane-hydrosilane copolymers at -75oC. Heating the compound with tetrabutoxydianilinocyclophosphazene at temperatures between 100oC-160oC may yield cyclophosphazene oligomers.{42}

Specifications

CAS 80-10-4
Density 1.204 g/mL (at 25°C)
Boiling Point 305°C
Molecular Formula C12H10Cl2Si
Refractive Index n20/D 1.578 (literature)
Linear Formula (C6H5)2 SiCl2
Quantity 100 g
MDL Number MFCD00000489
Synonym Diphenyldichlorosilane
Molecular Weight (g/mol) 253.2
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.