Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorododecylmethylsilane 95.0+%, TCI America™
SDP

Catalog No. D234925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D234925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D234925ML Supplier TCI America Supplier No. D234925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18407-07-3
Molecular Formula C13H28Cl2Si
Molecular Weight (g/mol) 283.352
MDL Number MFCD00018082
InChI Key ZAADUWCLWTWDSZ-UHFFFAOYSA-N
PubChem CID 87628
IUPAC Name dichloro-dodecyl-methylsilane
SMILES CCCCCCCCCCCC[Si](C)(Cl)Cl

Specifications

Boiling Point 127°C
Quantity 25 mL
UN Number 2987
Formula Weight 283.35
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorododecylmethylsilane
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.