Learn More
Abcam 2', 7'-Dichlorofluorescin diacetate, 25MG
Supplier:  Abcam AB27364025MG
Encompass_Preferred
MW 487.3 Da. Cell-permeable ROS probe. Reduced derivative of 2',7'-dichlorofluorescein. Can detect reactive oxygen species in live cells. Detects peroxidase activity (oxidative metabolism, intracellular regulation of ROS or oxidative burst). Upon enzyme activity, the highly fluorescent dye 2',7'-dichlorofluorescein is produced (Ex/Em = 495/529 nm).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-142