Learn More
Abcam 2',7'-Dichlorofluorescein di-ß-D-galactopyranoside (DCFDG), 25MG
Catalog No. 50996217
Encompass_Preferred
MW 725.48 g/mol, Purity >98%. Highly sensitive fluorescent substrate for measuring galactosidase and galactocerebrosidase activity. Since the pKa of the released fluorophore is significantly lower than comparable fluorophores, it can retain appreciably more fluorescence in the highly acidic environment of the lysosome than other similar fluorophores. Absorption prior to enzyme hydrolysis is 290 nm.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-217 Supplier Abcam Supplier No. AB27592025MG
MW 725.48 g/mol, Purity >98%. Highly sensitive fluorescent substrate for measuring galactosidase and galactocerebrosidase activity. Since the pKa of the released fluorophore is significantly lower than comparable fluorophores, it can retain appreciably more fluorescence in the highly acidic environment of the lysosome than other similar fluorophores. Absorption prior to enzyme hydrolysis is 290 nm.
The product is subject to the following: Abcam Restricted Use Statement