Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorofluorescein, 0.2% In 70% Isopropanol, Reagents

Catalog No. 01335430
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-430 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-430 Supplier Reagents Holdings Llc Supplier No. CD226500100C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dichlorofluorescein, 0.2% In 70% Isopropanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 7732-18-5 , 76-54-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym 2',7'-Dichloro-3,6-fluorandiol, 2,7-Dichlorofluorescein, 2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Boiling Point 82°C
Packaging Glass Bottle
Specific Gravity 0.86
Color Yellow/Green
Quantity 100 mL
Physical Form Liquid
Percent Purity 0.2%
Grade Reagent
Chemical Name or Material Dichlorofluorescein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.