Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorofluorescein TS, (U.S.P. Test Solution), Spectrum™ Chemical
SDP

Catalog No. 18602175 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-175 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-175 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D106500ML
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-106, 76-54-0

Spectrum™ Chemical Dichlorofluorescein TS, (U.S.P. Test Solution) is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 76-54-0 , 64-17-5 , 7732-18-5 , 1310-73-2
Molecular Formula C20H10Cl2O5
Molecular Weight (g/mol) 401.20
InChI Key VFNKZQNIXUFLBC-UHFFFAOYSA-N
IUPAC Name 2',7'-dichloro-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES OC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(O)C=C1O2

Specifications

CAS Max % 0.1%, 56.9%, 42.9%, 0.1%
UN Number UN1170
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 500 mL
Chemical Name or Material Dichlorofluorescein TS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.