Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichlorohexylmethylsilane 95.0+%, TCI America™
SDP

Catalog No. H085525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H085525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H085525ML Supplier TCI America Supplier No. H085525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14799-94-1
Molecular Formula C7H16Cl2Si
Molecular Weight (g/mol) 199.19
MDL Number MFCD00039295
InChI Key KKRMHVJQWMXYBZ-UHFFFAOYSA-N
Synonym Hexylmethyldichlorosilane
PubChem CID 84650
IUPAC Name dichloro(hexyl)methylsilane
SMILES CCCCCC[Si](C)(Cl)Cl

Specifications

Boiling Point 103°C
Quantity 25 mL
UN Number 2987
Formula Weight 199.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dichlorohexylmethylsilane
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.