Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dichloroiodomethane | 594-04-7 | 95.0% | CHCl2I | 10MG
SDP

Catalog No. 5000259945
Encompass_Preferred

Dichloroiodomethane (CHCl2I) is a trihalomethane research reagent used as an endogenous metabolite reference and analytical standard. It is supplied as a small-volume solution for laboratory research and requires cold, light-protected storage.

  • High purity, 95.0%.
  • Chemical formula CHCl2I.
  • Supplied in small vial sizes and as a 10 mM solution in ethanol.
  • Pale purple to purplish red liquid appearance.
  • Store at -20°C and protect from light.
  • Intended for research use only.

Catalog No. 50-002-59945 Supplier Medchemexpress LLC Supplier No. HY13364210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dichloroiodomethane (CHCl2I) is a trihalomethane research reagent used as an endogenous metabolite reference and analytical standard. It is supplied as a small-volume solution for laboratory research and requires cold, light-protected storage.

  • High purity, 95.0%.
  • Chemical formula CHCl2I.
  • Supplied in small vial sizes and as a 10 mM solution in ethanol.
  • Pale purple to purplish red liquid appearance.
  • Store at -20°C and protect from light.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.