Learn More
Medchemexpress LLC Dichloroiodomethane | 594-04-7 | MFCD30720198 | 95.0% | 210.83 g/mol | CHCl2I | 5 MG
SDP
Catalog No. 5000262546
Encompass_Preferred
Dichloroiodomethane (CAS 594-04-7) is an endogenous trihalomethane supplied as a small-volume laboratory reagent. It is provided as a colored liquid solution in ethanol at 95.0% purity and prepared to 10 mM concentration for convenient use. The material should be stored protected from light and handled according to supplied safety documentation.
Catalog No. 50-002-62546 Supplier Medchemexpress LLC Supplier No. HY1336425MG
Dichloroiodomethane (CAS 594-04-7) is an endogenous trihalomethane supplied as a small-volume laboratory reagent. It is provided as a colored liquid solution in ethanol at 95.0% purity and prepared to 10 mM concentration for convenient use. The material should be stored protected from light and handled according to supplied safety documentation.