Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloroisocyanuric acid sodium salt, 97% (dry wt.)

Catalog No. AAB2350414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2350414 25 g
AAB2350422 100 g
AAB2350436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2350414 Supplier Thermo Scientific Chemicals Supplier No. B2350414
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2893-78-9 | C3HCl2N3NaO3 | 220.949 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium dichloroisocyanurate can be used as a reagent for N-monochlorination and dehydrochlorination of amino esters, synthesis of antimicrobial polyurethane additives to control biofilm. It is used as a emergency disinfectant in drinking water, widely used cleansing agent. It is also a reagent for chlorination to detect ammonium via formation of colored zebra-bands in a detecting tube.

Solubility
Soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2893-78-9
Molecular Formula C3HCl2N3NaO3
Molecular Weight (g/mol) 220.949
MDL Number MFCD00006036
InChI Key UNWRHVZXVVTASG-UHFFFAOYSA-N
Synonym sodium dichlorocyanuric acid, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3-dichloro-, sodium salt
PubChem CID 86657659
IUPAC Name 1,3-dichloro-1,3,5-triazinane-2,4,6-trione;sodium
SMILES C1(=O)NC(=O)N(C(=O)N1Cl)Cl.[Na]

Specifications

Melting Point ∼250°C (decomposition)
Odor Chlorine-like
Quantity 25 g
UN Number UN2465
Beilstein 4056155
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 219.95
Percent Purity 97%
Assay (dry wt.), water <3%
Chemical Name or Material Dichloroisocyanuric acid sodium salt
Show More Show Less
Hazard Category H272-H302-H319-H335
Hazard Statement GHS H Statement
H272-H302-H319-H335
May intensify fire; oxidizer.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: DICHLOROISOCYANURIC ACID, SALTS
EINECSNumber 220-767-7
RTECSNumber XZ1900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.