Learn More
Sigma Aldrich Fine Chemicals Biosciences Dichloromethane HPLC Plus, for HPLC, GC, and residue analysis, >=99.9%, contains 50-150 ppm amylene as stabilizer | 75-09-2 | MFCD00000881 | 4L
Catalog No. NC9995998
Encompass
Dichloromethane HPLC Plus, for HPLC, GC, and residue analysis, >=99.9%, contains 50-150 ppm amylene as stabilizer | Purity: >=99.9% | Mol Wt: 84.93 | 75-09-2 | MFCD00000881 | 4L
This item is not returnable. View return policy
Catalog No. NC9995998 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 6504634L
 Restricted Item
Add to Cart
Add to Cart
Dichloromethane HPLC Plus, for HPLC, GC, and residue analysis, >=99.9%, contains 50-150 ppm amylene as stabilizer | Purity: >=99.9% | Mol Wt: 84.93 | 75-09-2 | MFCD00000881 | 4L