Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, Environmental Grade, 99.8+%, stab. with amylene

Catalog No. AA42006K2
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42006K2 1 L
AA42006M1 4 x 1 L
AA42006K7 4 L
AA42006M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AA42006K2 Supplier Thermo Scientific Chemicals Supplier No. 042006K2
Only null left
Add to Cart
This item is not returnable. View return policy

Suitable For Pesticide Residue Analysis | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Cl 10ppm, Halogenated residue 10ng/L heptachlorepoxide, Evaporation residue 3ppm, H2O 0.02%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Color Colorless
Appearance Colorless liquid
Density 1.325 g/cm3 at 20°C
Quantity 1 L
Ignition Point 605°C (1121°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.4244
Vapor Pressure 475 hPa (356mm Hg) at 20°C
Beilstein 1730800
Boiling Point 39°C to 40°C
Solubility Information Miscible with ethyl acetate,alcohol,hexanes,methanol,diethyl ether,n-octanol,acetone benzene,carbon tetrachloride,diethyl ether and chloroform. Immiscible with water.
Formula Weight 84.93
Grade Environmental
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Category H315-H319-H336-H350-H373
Precautionary Statement P201-P202-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 200-838-9
RTECSNumber PA8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.