Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.9%, for peptide synthesis, stabilized with amylene

Catalog No. p-3736183
Change view
Click to view available options
Quantity:
2.5 L
10 L
Packaging:
Glass Bottle
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC354800025 2.5 L Glass Bottle
AC354800100 10 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC354800025 Supplier Thermo Scientific Chemicals Supplier No. 354800025
Only null left
Add to Cart

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For peptide synthesis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2 , 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97.0°C
Density 1.3250g/mL
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.325
Assay Percent Range 99.9%
Linear Formula CH2Cl2
Viscosity 0.43 mPa.s (20°C)
Water 0.005% max.
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Merck Index 15, 6135
Boiling Point 39.0°C to 40.0°C
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Formula Weight 84.93
Percent Purity 99.9%
Chemical Name or Material Dichloromethane, Stabilized with Amylene
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement Use personal protective equipment as required. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.