Learn More
Sigma Aldrich Fine Chemicals Biosciences Dichloromethane anhydrous
Catalog No. 502898699
Encompass
Dichloromethane is a chlorinated organic solvent that is susceptible to degradation with time. This can be suppressed by adding amylene as a stabilizer.
Catalog No. 50-289-8699 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2709972L
Dichloromethane is a chlorinated organic solvent that is susceptible to degradation with time. This can be suppressed by adding amylene as a stabilizer.