Learn More
Sigma Aldrich Fine Chemicals Biosciences Dichloromethane anhydrous, contains 40-150 ppm amylene as stabilizer, ZerO2(R), >=99.8% | 75-09-2 | MFCD00000881 | 4X2ML
Catalog No. 501859761
Encompass
Dichloromethane anhydrous, contains 40-150 ppm amylene as stabilizer, ZerO2(R), >=99.8% | Purity: >=99.8% | Mol Wt: 84.93 | 75-09-2 | MFCD00000881 | 4X2ML
Catalog No. 50-185-9761 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 9006334X2ML
Dichloromethane anhydrous, contains 40-150 ppm amylene as stabilizer, ZerO2(R), >=99.8% | Purity: >=99.8% | Mol Wt: 84.93 | 75-09-2 | MFCD00000881 | 4X2ML