Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, Anhydrous, septum-sealed, for applications requiring low water content, contains amylene, >99.9%, Solstice

Catalog No. 6000486 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 100 mL
Packaging:
Septum-sealed Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-004-86 6 x 100 mL Septum-sealed Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-004-86 Supplier Solstice Supplier No. AS2990100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anhydrous, septum-sealed, for applications requiring low water content, contains amylene, >99.9%

  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

CAS Max % 100.0000%
Density 1.33 g/cm3
Quantity 6 x 100 mL
Packaging Septum-sealed Bottle
Physical Form anhydrous
UN Number UN1593
Beilstein 1730800
Boiling Point 40°C
Formula Weight 84.94
Percent Purity >99.9%
Chemical Name or Material Dichloromethane, Anhydrous, septum-sealed, for applications requiring low water content, contains amylene, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.