Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains cyclohexene , >99.9%, Solstice

Catalog No. 6000505 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
Packaging:
Returnable NOWPAK™ II
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-005-05 20 L Returnable NOWPAK™ II
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-005-05 Supplier Solstice Supplier No. NS30020
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Absorbance 0.1 AU max. at 240 nm, 0.01 AU max. at 250 nm
CAS Max % 100.0000%
Density 1.33 g/cm3
Quantity 20 L
Packaging Returnable NOWPAK™ II
Linear Formula H2O
Water 0.01% max.
UN Number UN1593
Beilstein 1730800
Boiling Point 40°C
Solubility Information 160% in Water at 20°C
Formula Weight 84.94
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Dichloromethane, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains cyclohexene , >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.