Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, BioSyn™, for biosynthesis and applications requiring low water content, contains cyclohexene , >99.9%, Solstice

Catalog No. 60048423 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-423 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-048-423 Supplier Solstice Supplier No. BB3014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym Methylene Chloride, DCM
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

CAS Max % 100.00
Density 1.33 g/cm3
Quantity 4 x 4 L
Flash Point does not flash
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 40°C
Grade Low Water
Hazard Category Warning
Hazard Statement H315+H319+H336+H351
Precautionary Statement P280+P271+P264+P261+P202+P201
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.