Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, Biotech Grade, 99.9%,Solstice

Catalog No. 60046981 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-981 1 L Glass Bottle
60-046-982 2 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-981 Supplier Solstice Supplier No. 4944531L
Add to Cart
Add to Cart

Biotech Grade, 99.9%, contains 40-150 ppm amylene as stabilizer

  • Solstice developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Solstice continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Density 1.325g/mL at 25°C
Quantity 1 L
Explosion Limit 0.22
Ignition Point 1223°F
Odor Sweet
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 24.45psi (55°C), 6.83psi (20°C)
UN Number UN1593
Beilstein 1730800
Boiling Point 39°C to 41°C
Formula Weight 84.93g/mol
Percent Purity 99.90%
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338
DOTInformation 6.1/PGIII
RTECSNumber 200-838-9
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.