Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane ChromAR, Macron Fine Chemicals™

Catalog No. MK48791 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass
SAFEMOR
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-4879-1 1 L Glass
MK-4879-4 4 L Glass
MK-H077-4 4 L SAFEMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MK-4879-1 Supplier Avantor Supplier No. 487906
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Quantity 1 L
Packaging Glass
Formula Weight 84.93
Chemical Name or Material Dichloromethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.