Learn More
Sigma Aldrich Fine Chemicals Biosciences Dichloromethane contains 40-150 ppm amylene as stabilizer, ACS reagent, >=99.5% | 75-09-2 | MFCD00000881 | 4X4L
Catalog No. 501859769
Encompass
Dichloromethane contains 40-150 ppm amylene as stabilizer, ACS reagent, >=99.5% | Purity: >=99.5% | Mol Wt: 84.93 | 75-09-2 | MFCD00000881 | 4X4L
Catalog No. 50-185-9769 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D651004X4L
Dichloromethane contains 40-150 ppm amylene as stabilizer, ACS reagent, >=99.5% | Purity: >=99.5% | Mol Wt: 84.93 | 75-09-2 | MFCD00000881 | 4X4L