Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane-d{2}, 99.9% (Isotopic)

Catalog No. AA42335SA
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42335SA 2 Ea.
AA42335KV 10 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42335SA Supplier Thermo Scientific Chemicals Supplier No. 042335SA
Add to Cart

Acts as a solvent in Nuclear magnetic resonance (NMR) spectroscopy. | CAS: 1665-00-5

Dichloromethane-d{2} acts as a solvent in Nuclear magnetic resonance (NMR) spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
MDL Number MFCD00000882
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d

Specifications

Density 1.35
Packaging Prescored ampoules, 0.75mL/ampoule
Quantity 2 Ea.
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohol,ether and dimethyl formamide.
Formula Weight 86.95
Percent Purity 99.9%
Physical Form Liquid
Chemical Name or Material Dichloromethane-d2
Hazard Category H315-H319-H336-H351
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 216-776-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.