Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane-d{2}, 99.9% (Isotopic)

Catalog No. AA3650006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3650006 5 g
AA3650003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3650006 Supplier Thermo Scientific Chemicals Supplier No. 03650006
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol

In Anion-templated assembly of [2] rotaxanes, the addition of tetrabutylammonium (TBA) halide, dihydrogen phosphate and hydrogen sulfate salts to dichloromethane-d 2 solutions of both derivatives resulted in significant downfield shift perturbations of the respective compounds' aryl b and amide c protons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 86.939
MDL Number MFCD00000882
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
PubChem CID 160586
IUPAC Name dichloro(dideuterio)methane
SMILES C(Cl)Cl

Specifications

Physical Form Liquid
Sensitivity Moisture
Quantity 5 g
Density 1.35 g/mL
Odor Sweet
UN Number UN1593
Solubility Information Insoluble in water. Soluble in acetone,chloroform,tetrachloromethane,dimethylfomrarnide.
Formula Weight 86.95
Percent Purity 99.9%
Chemical Name or Material Dichloromethane-d2, Isotopic
Hazard Category H315-H319-H336-H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DICHLOROMETHANE
EINECSNumber 216-776-0
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.