Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane - D2, deuteration degree min. 99.96% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Catalog No. M1042000010 Shop All MilliporeSigma Products
Change view
Click to view available options
Glass ampoule; 10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
M1042000010 Glass ampoule; 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1042000010 Supplier MilliporeSigma Supplier No. 1.04200.0010

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mL
Physical Form Liquid
Packaging Glass Ampule
Vapor Pressure ca. 450 hPa at 20°C (68°F)
Odor Sweet
Color Colorless
CAS 1665-00-5
Assay Percent Range 99.96% min.
Molecular Formula CH2Cl2
Synonym dichloromethane-d2, methylene chloride-d2, dichloro 2h2 methane, methane-d2, dichloro, cd2cl2, deuterated dichloromethane, dichloro dideuterio methane, dideuteromethylene chloride, dichloromethane-d2, 99.9 atom % d, dichloromethane-d', 99.96 atom % d
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
SMILES C(Cl)Cl
IUPAC Name dichloro(dideuterio)methane
Molecular Weight (g/mol) 86.939
PubChem CID 160586
Formula Weight 86.95
Chemical Name or Material Dichloromethane - D2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.