Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane-d2, for NMR, 99.5 atom % D

Catalog No. AC176110100
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
50 mL
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176110100 10 mL Glass Bottle
AC176110050 5 mL Glass Bottle
AC176110250 25 mL Glass Bottle
AC176110500 50 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC176110100 Supplier Thermo Scientific Chemicals Supplier No. 176110100
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1665-00-5 | CH2Cl2 | 86.939 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
MDL Number MFCD00000882
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
Synonym dichloromethane-d2, 99.9 atom % d, dichloro dideuterio methane, cd2cl2, dichloromethane-d', 99.96 atom % d, dichloro 2h2 methane, dideuteromethylene chloride, methylene chloride-d2, methane-d2, dichloro, dichloromethane-d2, deuterated dichloromethane
PubChem CID 160586

Specifications

Physical Form Liquid
Color Colorless
Deuterium (D) 99.60% min.
Packaging Glass Bottle
Water 0.04% max. (Coulometric)
Quantity 10 mL
Density 1.3600 g/mL
Refractive Index 1.4218
Melting Point -97.0°C
Boiling Point 40.0°C
Linear Formula CD2Cl2
Beilstein 01, IV, 39
SMILES [2H]C([2H])(Cl)Cl
IUPAC Name dichloro(²H₂)methane
Molecular Weight (g/mol) 86.94
Formula Weight 86.95
Grade NMR
Chemical Name or Material Dichloromethane-d2, 99.6+ atom % D
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:

Suspected of causing cancer.
Precautionary Statement GHS P Statement:

Use personal protective equipment as required.



WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-776-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.