Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane-d2, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D

Catalog No. AC464030075
Change view
Click to view available options
Quantity:
7.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464030075 7.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC464030075 Supplier Thermo Scientific Chemicals Supplier No. 464030075
 Restricted Item
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1665-00-5 | CH2Cl2 | 86.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1665-00-5
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 86.94
InChI Key YMWUJEATGCHHMB-DICFDUPASA-N
IUPAC Name dichloro(²H₂)methane
SMILES [2H]C([2H])(Cl)Cl

Specifications

Refractive Index 1.4218
Density 1.3600 g/mL
Quantity 7.5 mL
Specific Gravity 1.36
Melting Point -97.0°C
Boiling Point 40.0°C
Linear Formula CD2Cl2
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in alchol, ether and dmf
Formula Weight 86.95
Grade NMR
Chemical Name or Material Dichloromethane-d2
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of causing cancer.

Causes skin irritation.

Causes serious eye irritation.

May cause drowsiness or dizziness.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 216-776-0

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.