Learn More
Sigma Aldrich Fine Chemicals Biosciences Dichloromethane suitable for HPLC, >=99.9%, contains 40-150 ppm amylene as stabilizer | 75-09-2 | MFCD00000881 | 4L
Catalog No. 501859745
Encompass
Dichloromethane suitable for HPLC, >=99.9%, contains 40-150 ppm amylene as stabilizer | Purity: >=99.9% | Mol Wt: 84.93 | 75-09-2 | MFCD00000881 | 4L
Catalog No. 50-185-9745 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4392234L
Dichloromethane suitable for HPLC, >=99.9%, contains 40-150 ppm amylene as stabilizer | Purity: >=99.9% | Mol Wt: 84.93 | 75-09-2 | MFCD00000881 | 4L