Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, For pesticide residue analysis, Solstice

Catalog No. 60046348 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-348 4 x 2.5 L
60-004-99 1 L
60-005-02 4 x 4 L
60-005-00 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 60-046-348 Supplier Solstice Supplier No. 344884X2.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C (literature)
CAS Min % ≥99.0000%
CAS Max % ≥99.0000%
Density 1.325 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Packaging Glass Bottle
Linear Formula CH2Cl2
UN Number UN1593
Beilstein 1730800
Boiling Point 39°C to 41°C
Formula Weight 84.93
Percent Purity >99.8%
Grade Pesticide
Chemical Name or Material Dichloromethane, For pesticide residue analysis
Show More Show Less
ShelfLife 720 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Dichloromethane
EINECSNumber 200-838-9
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.