Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane HPLC, Lab Chem

Catalog No. LC135824 Shop All LabChem Products
Change view
Click to view available options
Appearance:
Colorless Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Appearance
LC135824 Colorless Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC135824 Supplier LabChem, Inc. Supplier No. LC135824
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Dichloromethane HPLC Solvent, Safety: Toxic

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Color Colorless
Appearance Colorless Liquid
Density 1.3266 g/cm3
Quantity 4 L
Odor Mild, sweet
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation < 3 ppm
Specific Gravity 1.3266
Titratable Acid <0.0003meq/g
Linear Formula CH2Cl2
Vapor Pressure 6.83 psi at 20 °C
UN Number UN1593
Boiling Point 103.3 °F
Formula Weight 84.93
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Dichloromethane HPLC
Show More Show Less
Hazard Category Eye Irritation
Hazard Statement Suspected cause of cancer
ShelfLife 3 Years
DOTInformation UN1593
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.