Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, Puriss., meets analytical spec. of Ph. Eur., NF, ≥99% (GC), Solstice

Catalog No. 60045589 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
35 kg
4 x 2.5 L
4 x 5 L
6 x 1 L
250 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Stainless Steel Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-589 6 x 1 L Glass Bottle
60-045-583 2.5 L Glass Bottle
60-045-588 5 L Aluminum Bottle
60-045-586 4 x 2.5 L Glass Bottle
60-045-587 4 x 5 L Aluminum Bottle
60-045-585 35 kg Stainless Steel Can
60-045-584 250 kg Metal Drum
60-045-582 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 60-045-589 Supplier Solstice Supplier No. 242336X1L
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss., meets analytical specification of Ph. Eur., NF,≥99% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Density 1.318 to 1.322g/mL at 25°C, 1.325g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.22
Ignition Point 1223°F
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 24.45psi (55°C), 6.83psi (20°C)
UN Number UN1593
Beilstein 1730800
Boiling Point 39°C to 41°C
Formula Weight 84.93g/mol
Percent Purity ≥99% (GC)
Grade puriss.
Chemical Name or Material Dichloromethane, Meets analytical specification of Ph.Eur.NF
Show More Show Less
Hazard Statement P261-P281-P305 + P351 + P338
Precautionary Statement H315-H319-H335-H336-H351-H373
DOTInformation 6.1/PGIII
EINECSNumber 200-838-9
RTECSNumber PA8050000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.