Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane solution contains 10% (v/v) Methanol, MilliporeSigma™ Supelco™

Catalog No. 111017972 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7972 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7972 Supplier MilliporeSigma Supelco Supplier No. 7238514X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The product is a 10%v/v solution of dichloromethane in methanol. Dichloromethane (DCM) is a chlorinated organic solvent that has C2V symmetry, low boiling point, high density and is immiscible in water. A study reports the dielectric characteristics of DCM over a wide frequency range.

Food and Beverages
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00000881
Synonym Methylene chloride solution

Specifications

Density 1.269 g/mL (at 25°C)
CAS 75-09-2
Quantity 4 x 4 L
Packaging Glass Bottle
Linear Formula CH2 Cl2
Formula Weight 84.93
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.