Learn More
Sigma Aldrich Fine Chemicals Biosciences Dichloromethane suitable f
Catalog No. 502903089
Encompass
Dichloromethane has been tested as a solvent medium for dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml. Role of the quantity of TiO2 loading on activated carbon support on dichloromethane photodecomposition has been investigated.
Catalog No. 50-290-3089 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348564X4L
Dichloromethane has been tested as a solvent medium for dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml. Role of the quantity of TiO2 loading on activated carbon support on dichloromethane photodecomposition has been investigated.