Learn More
Sigma Aldrich Fine Chemicals Biosciences Dichloromethane suitable f1L
Catalog No. 502897310
Encompass
Dichloromethane has been tested as a solvent medium for dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml. Role of the quantity of TiO2 loading on activated carbon support on dichloromethane photodecomposition has been investigated.
Catalog No. 50-289-7310 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 348561L
Dichloromethane has been tested as a solvent medium for dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml. Role of the quantity of TiO2 loading on activated carbon support on dichloromethane photodecomposition has been investigated.